Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GLRX5 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA524956
Description
GLRX5 Polyclonal Antibody for Western Blot, Flow
GLRX5 is monothiol glutaredoxin involved in the biogenesis of iron-sulfur clusters. GLRX5 required for normal iron homeostasis. GLRX5 is required for normal regulation of hemoglobin synthesis by the iron-sulfur protein ACO1.
Specifications
GLRX5 | |
Polyclonal | |
PBS with 0.09% sodium azide; pH 7.4 | |
Q86SX6 | |
GLRX5 | |
KLH conjugated synthetic peptide between 129-157 amino acids from the C-terminal region of human GLRX5. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Flow Cytometry, Western Blot | |
Unconjugated | |
GLRX5 | |
2310004O13Rik; 2900070E19Rik; AU020725; C14orf87; FLB4739; GLRX5; glutaredoxin 5; glutaredoxin 5 homolog; glutaredoxin 5 homolog (S. cerevisiae); glutaredoxin-related protein 5, mitochondrial; GRX5; Monothiol glutaredoxin-5; PR01238; PRO1238; PRSA; RGD1308383; SIDBA3; SPAHGC | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
51218 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction