Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GLS Rabbit anti-Human, Polyclonal, MilliporeSigma™

Catalog No. ABN1466MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABN1466MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABN1466MI Supplier MilliporeSigma Supplier No. ABN1466
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-GLS is a highly specific rabbit polyclonal antibody that targets Glutaminase kidney isoform, mitochondrial, and has been tested in Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GLS
Applications Western Blot
Classification Polyclonal
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Gene Symbols GLS,GLS1,KIAA0838
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to 16 amino acids from the C-terminal region.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) NP_055720
Test Specificity This polyclonal antibody detects the Glutaminase kidney isoform, mitochondrial, in human. Also detects GLS in the neural cortex. It targets an epitope within 16 amino acids fromt he C-terminal region.
Target Species Human
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.