Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GLS2 Monoclonal Antibody (OTI4C10)
GREENER_CHOICE

Catalog No. PIMA527167
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA527167 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA527167 Supplier Invitrogen™ Supplier No. MA527167
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

GLS2 Monoclonal Antibody for Western Blot, IHC (P)

The protein encoded by this gene is a mitochondrial phosphate-activated glutaminase that catalyzes the hydrolysis of glutamine to stoichiometric amounts of glutamate and ammonia. This protein is functionally similar to the kidney glutaminase but is a little smaller in size. Originally thought to be liver-specific, this protein has been found in other tissues as well. At least one transcribed pseudogene has been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GLS2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI4C10
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Gls2
Gene Accession No. Q9UI32
Gene Alias A330074B06Rik; AI195532; breast cell glutaminase; Ga; GLS; GLS2; glutaminase 2; glutaminase 2 (liver, mitochondrial); glutaminase GA; glutaminase I; glutaminase liver isoform, mitochondrial; hLGA; Kiaa4146; L hL L-glutaminase; L hLGA; LGA; L-glutaminase; L-glutamine amidohydrolase; liver mitochondrial glutaminase; phosphate-activated glutaminase; phosphate-dependent glutaminase
Gene Symbols Gls2
Host Species Mouse
Immunogen Full length human recombinant protein of GLS2 produced in E.coli.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27165
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.