Learn More
Invitrogen™ GLT-1 Polyclonal Antibody

Description
Positive Samples: MCF7, HepG2, SW480, Mouse liver, Rat liver; Cellular Location: Membrane, Multi-pass membrane protein Immunogen sequence: LDRMRTSVNV VGDSFGAGIV YHLSKSELDT IDSQHRVHED IEMTKTQSIY DDMKNHRESN SNQCVYAAHN SVIVDECKVT LAANGKSADC SVEEEPWKRE K.
Specifications
Specifications
| Antigen | GLT-1 |
| Applications | ELISA, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.18 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | SLC1A2 |
| Gene Accession No. | P43004, P43006 |
| Gene Alias | 1700091C19Rik; 2900019G14Rik; AI159670; Eaat2; EAAT2 variant mEAAT2/5UT5; excitatory amino acid transporter 2; excitotoxic amino acid transporter 2; glial high affinity glutamate transporter; Glt; GLT1; GLT-1; GLT1a; GLT1b; GLT1c; glutamate transporter 1c; glutamate/aspartate transporter II; GLUT-R; HBGT; high affinity glucose transporter; MGLT1; OTTHUMP00000165209; OTTHUMP00000165210; SLC1A2; Sodium-dependent glutamate/aspartate transporter 2; solute carrier family 1 (glial high affinity glutamate transporter), member 2; solute carrier family 1 member 2; solute carrier family 1, member 2 |
| Gene Symbols | SLC1A2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.