Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GLT-1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5115151
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5115151 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5115151 Supplier Invitrogen™ Supplier No. PA5115151
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total EAAT2.

Excitatory amino acid transporter 2 (EAAT2) is a member of a family of solute transporter proteins. The membrane-bound protein is the principal transporter that clears the excitatory neurotransmitter glutamate from the extracellular space at synapses in the central nervous system. Glutamate clearance is necessary for proper synaptic activation and to prevent neuronal damage from excessive activation of glutamate receptors. Mutations in and decreased expression of this protein are associated with amyotrophic lateral sclerosis (ALS.) Alternatively spliced transcript variants of this gene have been identified.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GLT-1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene SLC1A2
Gene Accession No. P31596, P43004, P43006
Gene Alias 1700091C19Rik; 2900019G14Rik; AI159670; Eaat2; EAAT2 variant mEAAT2/5UT5; excitatory amino acid transporter 2; excitotoxic amino acid transporter 2; glial high affinity glutamate transporter; Glt; GLT1; GLT-1; GLT1a; GLT1b; GLT1c; glutamate transporter 1c; glutamate/aspartate transporter II; GLUT-R; HBGT; high affinity glucose transporter; MGLT1; OTTHUMP00000165209; OTTHUMP00000165210; SLC1A2; Sodium-dependent glutamate/aspartate transporter 2; solute carrier family 1 (glial high affinity glutamate transporter), member 2; solute carrier family 1 member 2; solute carrier family 1, member 2
Gene Symbols SLC1A2
Host Species Rabbit
Immunogen A synthesized peptide derived from human SLC1A2(Accession P43004), corresponding to amino acid residues M125-L175.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20511, 29482, 6506
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.