Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GLT-1 Recombinant Rabbit Monoclonal Antibody (PS01-62)
GREENER_CHOICE

Catalog No. FEMA550813
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEMA550813 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEMA550813 Supplier Invitrogen™ Supplier No. MA550813
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Positive control: Mouse brain tissue lysate, rat brain tissue lysate, mouse cerebellum tissue lysate, mouse brain tissue, mouse cerebellum tissue, rat cerebellum tissue. Predicted band size: 62 kDa Subcellular Location: Cell membrane.

Excitatory amino acid transporter 2 (EAAT2) is a member of a family of solute transporter proteins. The membrane-bound protein is the principal transporter that clears the excitatory neurotransmitter glutamate from the extracellular space at synapses in the central nervous system. Glutamate clearance is necessary for proper synaptic activation and to prevent neuronal damage from excessive activation of glutamate receptors. Mutations in and decreased expression of this protein are associated with amyotrophic lateral sclerosis (ALS.) Alternatively spliced transcript variants of this gene have been identified.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GLT-1
Applications Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot
Classification Recombinant Monoclonal
Clone PS01-62
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
Gene SLC1A2
Gene Accession No. P31596, P43004, P43006
Gene Alias 1700091C19Rik; 2900019G14Rik; AI159670; Eaat2; EAAT2 variant mEAAT2/5UT5; excitatory amino acid transporter 2; excitotoxic amino acid transporter 2; glial high affinity glutamate transporter; Glt; GLT1; GLT-1; GLT1a; GLT1b; GLT1c; glutamate transporter 1c; glutamate/aspartate transporter II; GLUT-R; HBGT; high affinity glucose transporter; MGLT1; OTTHUMP00000165209; OTTHUMP00000165210; SLC1A2; Sodium-dependent glutamate/aspartate transporter 2; solute carrier family 1 (glial high affinity glutamate transporter), member 2; solute carrier family 1 member 2; solute carrier family 1, member 2
Gene Symbols SLC1A2
Host Species Rabbit
Immunogen Synthetic peptide within Human GLT-1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20511, 29482, 6506
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.