Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GLT25D1 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501982847
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-2847 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-2847 Supplier Bioss Supplier No. BS13378R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifications

Antigen GLT25D1
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene COLGALT1
Gene Accession No. Q8NBJ5
Gene Alias 2810024B22Rik; ColGalT 1; Colgalt1; Collagen beta(1-O)galactosyltransferase 1; Glt25d1; glycosyltransferase 25 domain containing 1; Glycosyltransferase 25 family member 1; Hydroxylysine galactosyltransferase 1; Procollagen galactosyltransferase; Procollagen galactosyltransferase 1; PSEC0241; RGD1309002
Gene Symbols COLGALT1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human GLT25D1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79709
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.