Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GLT6D1 Antibody, Novus Biologicals™
SDP

Catalog No. NBP17931020 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP17931020 20 μL
NBP179310 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP17931020 Supplier Novus Biologicals Supplier No. NBP17931020UL

Rabbit Polyclonal Antibody

GLT6D1 Polyclonal specifically detects GLT6D1 in Human samples. It is validated for Western Blot.

Specifications

Antigen GLT6D1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:1000
Formulation PBS and 2% Sucrose with 0.09% Sodium Azide
Gene Accession No. NP_892019
Gene Alias galactosyltransferase family 6 domain containing 1, galactosyltransferase family 6 domain-containing 1, glycosyltransferase 6 domain containing 1, glycosyltransferase 6 domain-containing protein 1, GT6M7
Gene Symbols GLT6D1
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human GLT6D1The immunogen for this antibody is GLT6D1. Peptide sequence FGVETLGPLVAQLHAWWYFRNTKNFPYERRPTSAACIPFGQGDFYYGNLM.
Purification Method Affinity Purified
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 360203
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.