Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GLT8D1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5144687
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5144687 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5144687 Supplier Invitrogen™ Supplier No. PA5144687
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VPNALRHAVD GRQEEIPVVI AASEDRLGGA IAAINSIQHN TRSNVIFYIV TLNNTADHLR SWLNSDSLKS IRYKIVNFDP KLLEGKVKED PDQGESMKPL TFARFYLPIL VPSAKKAIYM DDDVIVQG Antigen sequence identity: Mouse 87%; Rat 88%.

This gene encodes a member of the glycosyltransferase family. The specific function of this protein has not been determined. Alternative splicing results in multiple transcript variants of this gene [provided by RefSeq, May 2013].
TRUSTED_SUSTAINABILITY

Specifications

Antigen GLT8D1
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Glt8d1
Gene Accession No. Q68CQ7
Gene Alias 2410004H05Rik; 5430414N14Rik; AD-017; AI450005; Da2-24; GALA4A; Glt8d1; glycosyltransferase 8 domain containing 1; glycosyltransferase 8 domain-containing protein 1; glycosyltransferase AD-017; MSTP137; MSTP139; UNQ572/PRO1134
Gene Symbols Glt8d1
Host Species Rabbit
Immunogen Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range: 51-178. Recombinant protein control fragment (Product #RP-89987).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55830
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Format Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.