Learn More
Invitrogen™ GLT8D1 Polyclonal Antibody

Description
GLT8D1 Polyclonal Antibody for Western Blot
Specifications
Specifications
| Antigen | GLT8D1 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | Glt8d1 |
| Gene Accession No. | Q68CQ7 |
| Gene Alias | 2410004H05Rik; 5430414N14Rik; AD-017; AI450005; Da2-24; GALA4A; Glt8d1; glycosyltransferase 8 domain containing 1; glycosyltransferase 8 domain-containing protein 1; glycosyltransferase AD-017; MSTP137; MSTP139; UNQ572/PRO1134 |
| Gene Symbols | Glt8d1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.