Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GLT8D1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5113796
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5113796 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5113796 Supplier Invitrogen™ Supplier No. PA5113796
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LLIVFYQQHS TIDPMWNVRH LGSSAGKRYS PQFVKAAKLL HWNGHLKPWG For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.

This gene encodes a member of the glycosyltransferase family. The specific function of this protein has not been determined. Alternative splicing results in multiple transcript variants of this gene [provided by RefSeq, May 2013].
TRUSTED_SUSTAINABILITY

Specifications

Antigen GLT8D1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene Glt8d1
Gene Accession No. Q68CQ7
Gene Alias 2410004H05Rik; 5430414N14Rik; AD-017; AI450005; Da2-24; GALA4A; Glt8d1; glycosyltransferase 8 domain containing 1; glycosyltransferase 8 domain-containing protein 1; glycosyltransferase AD-017; MSTP137; MSTP139; UNQ572/PRO1134
Gene Symbols Glt8d1
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human GLT8D1.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55830
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.