Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GLT8D2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$322.50 - $534.50
Specifications
Antigen | GLT8D2 |
---|---|
Dilution | Immunocytochemistry/ Immunofluorescence 0.25-2 ug/mL |
Applications | Immunocytochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
GLT8D2 Polyclonal antibody specifically detects GLT8D2 in Human samples. It is validated for Immunocytochemistry/ ImmunofluorescenceSpecifications
GLT8D2 | |
Immunocytochemistry | |
Unconjugated | |
Rabbit | |
Human | |
EC 2.4.1, EC 2.4.1.-, FLJ31494, GALA4A, glycosyltransferase 8 domain containing 2, glycosyltransferase 8 domain-containing protein 2, gycosyltransferase | |
This antibody was developed against Recombinant Protein corresponding to amino acids: MGYLDYRKKAIKDLGISPSTCSFNPGVIVANMTEWKHQRITKQLEKWMQKNVEENLYSSSLGGGVATSPMLIVFHGKYSTINPLWHIRHLGWNPDARYSEHFLQEAKLLH | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Immunocytochemistry/ Immunofluorescence 0.25-2 ug/mL | |
Polyclonal | |
Purified | |
RUO | |
PBS, pH 7.2, 40% glycerol | |
83468 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title