Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GLT8D2 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501988447
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-8447 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-8447 Supplier Bioss Supplier No. BS8302R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifications

Antigen GLT8D2
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene GLT8D2
Gene Accession No. Q9H1C3
Gene Alias GALA4A; GLT8D2; glycosyltransferase 8 domain containing 2; Glycosyltransferase 8 domain-containing protein 2; gycosyltransferase; UNQ1901/PRO4347
Gene Symbols GLT8D2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human GLT8D2.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 83468
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.