Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GLT8D4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA564369
Description
Immunogen sequence: MNLTRIRSTQ FKNSMIPTGL AWEDMLYPLY QKYKNAITWG DQ Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 93%.
Glycosyltransferase which elongates the O-linked glucose attached to EGF-like repeats in the extracellular domain of Notch proteins by catalyzing the addition of xylose.
Specifications
GLT8D4 | |
Polyclonal | |
Unconjugated | |
GXYLT2 | |
242n2; GLT8D4; Glucoside xylosyltransferase 2; glycosyltransferase 8 domain containing 4; glycosyltransferase 8 domain-containing protein 4; GXYLT2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
727936 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
A0PJZ3 | |
GXYLT2 | |
Recombinant protein corresponding to Human GLT8D4. Recombinant protein control fragment (Product #RP-103823). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction