Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GLTP Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5119156
This item is not returnable.
View return policy
Description
GLTP Polyclonal Antibody for Western Blot, IHC (P)
Accelerates the intermembrane transfer of various glycolipids. Catalyzes the transfer of various glycosphingolipids between membranes but does not catalyze the transfer of phospholipids. May be involved in the intracellular translocation of glucosylceramides.
Specifications
GLTP | |
Polyclonal | |
Unconjugated | |
GLTP | |
1110001F24Rik; C76925; C77564; Gltp; Glycolipid transfer protein; RGD1564442; truncated glycolipid transfer protein | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
396765, 51228, 56356 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
P68266, Q9JL62, Q9NZD2 | |
GLTP | |
Recombinant protein Glycolipid Transfer Protein. The antigen corresponds to amino acid range 2-209 of the target protein. | |
200 μL | |
Primary | |
Human, Mouse, Pig | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction