Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GLTP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563306
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563306 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563306 Supplier Invitrogen™ Supplier No. PA563306
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ALLAEHLLKP LPADKQIETG PFLEAVSHLP PFFDCLGSPV FTPIKADISG Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 96%.

Accelerates the intermembrane transfer of various glycolipids. Catalyzes the transfer of various glycosphingolipids between membranes but does not catalyze the transfer of phospholipids. May be involved in the intracellular translocation of glucosylceramides.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GLTP
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene GLTP
Gene Accession No. Q9NZD2
Gene Alias 1110001F24Rik; C76925; C77564; Gltp; Glycolipid transfer protein; RGD1564442; truncated glycolipid transfer protein
Gene Symbols GLTP
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human GLTP. Recombinant protein control fragment (Product #RP-101667).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51228
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.