Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GLTPD2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
Antigen | GLTPD2 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
GLTPD2 Polyclonal specifically detects GLTPD2 in Human samples. It is validated for Western Blot.Specifications
GLTPD2 | |
Polyclonal | |
Rabbit | |
Human | |
glycolipid transfer protein domain containing 2, glycolipid transfer protein domain-containing protein 2 | |
GLTPD2 | |
IgG |
Western Blot | |
Unconjugated | |
RUO | |
NP_001014985 | |
388323 | |
Synthetic peptide directed towards the N terminal of human LOC388323. Peptide sequence GARSGCGPRAQPCVPGETAPFQVRQESGTLEAPERKQPPCLGPRGMLGRM. | |
Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title