Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GLTSCR1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA570729
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA570729 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA570729 Supplier Invitrogen™ Supplier No. PA570729
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Human: 100%; Mouse: 100%; Rat: 100%.

Component of SWI/SNF chromatin remodeling subcomplex GBAF that carries out key enzymatic activities, changing chromatin structure by altering DNA-histone contacts within a nucleosome in an ATP-dependent manner (PubMed:29374058). May play a role in BRD4-mediated gene transcription (PubMed:21555454). [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen GLTSCR1
Applications Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot, Surface Plasmon Resonance
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene BICRA
Gene Accession No. F8VPZ9, Q9NZM4
Gene Alias BICRA; BRD4 interacting chromatin remodeling complex associated protein; BRD4 interacting chromatin remodelling complex associated protein; BRD4-interacting chromatin-remodeling complex-associated protein; glioma tumor suppressor candidate region gene 1; glioma tumor suppressor candidate region gene 1 protein; glioma tumor suppressor candidate region gene 1 protein; LOW QUALITY PROTEIN: BRD4-interacting chromatin-remodeling complex-associated protein; GLTSCR1
Gene Symbols BICRA
Host Species Rabbit
Immunogen synthetic peptide directed towards the C-terminal region of mouse GLTSCR1.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 243842, 29998
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.