Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GLTSCR2 Antibody, Novus Biologicals™

Goat Polyclonal Antibody
Supplier: Novus Biologicals NBP106061
Description
GLTSCR2 Polyclonal specifically detects GLTSCR2 in Human, Mouse, Rat, Bovine, Canine samples. It is validated for Peptide ELISA.Specifications
GLTSCR2 | |
Polyclonal | |
Unconjugated | |
Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
glioma tumor suppressor candidate region gene 2, glioma tumor suppressor candidate region gene 2 protein, p60, PICT1, PICT-1, protein interacting with carboxyl terminus 1 | |
Goat | |
Affinity Purified | |
RUO | |
Primary | |
This antibody is expected to recognize a sequence corresponding to aa 38-51 of human PICT1 protein. | |
Store at -20C. Avoid freeze-thaw cycles. |
ELISA | |
0.5 mg/mL | |
Peptide ELISA Detection limit 1:32000 | |
NP_056525.1 | |
GLTSCR2 | |
Peptide with sequence CRRGPRNKKRGWRRL corresponding to internal region according to NP_056525.2. | |
0.1 mg | |
Cancer | |
29997 | |
Human, Mouse, Rat, Bovine, Canine | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction