Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GLTSCR2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553888
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553888 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553888 Supplier Invitrogen™ Supplier No. PA553888
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: REAEADKPRR LGRLKYQAPD IDVQLSSELT DSLRTLKPEG NILRDRFKSF QRRNMIEPRE RAKFKRKYKV KLVEKRAFRE IQL Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 95%.

Nucleolar protein which is involved in the integration of the 5S RNP into the ribosomal large subunit during ribosome biogenesis. In ribosome biogenesis, may also play a role in rRNA transcription. Also functions as a nucleolar sensor that regulates the activation of p53/TP53 in response to ribosome biogenesis perturbation, DNA damage and other stress conditions. DNA damage or perturbation of ribosome biogenesis disrupt the interaction between NOP53 and RPL11 allowing RPL11 transport to the nucleoplasm where it can inhibit MDM2 and allow p53/TP53 activation. It may also positively regulate the function of p53/TP53 in cell cycle arrest and apoptosis through direct interaction, preventing its MDM2-dependent ubiquitin-mediated proteasomal degradation. Originally identified as a tumor suppressor, it may also play a role in cell proliferation and apoptosis by positively regulating the stability of PTEN, thereby antagonizing the PI3K-AKT/PKB signaling pathway. May also inhibit cell proliferation and increase apoptosis through its interaction with NF2. May negatively regulate NPM1 by regulating its nucleoplasmic localization, oligomerization and ubiquitin-mediated proteasomal degradation. Thereby, may prevent NPM1 interaction with MYC and negatively regulate transcription mediated by the MYC-NPM1 complex. May also regulate cellular aerobic respiration. In the cellular response to viral infection, may play a role in the attenuation of interferon-beta through the inhibition of DDX58/RIG-1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GLTSCR2
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene NOP53
Gene Accession No. Q9NZM5
Gene Alias 5330430H08Rik; 9430097C02Rik; AU041936; AW536441; glioma tumor suppressor candidate region gene 2; glioma tumor suppressor candidate region gene 2 protein; GLT; GLTSCR2; LOW QUALITY PROTEIN: glioma tumor suppressor candidate region gene 2 protein; NOP53; NOP53 ribosome biogenesis factor; p60; PICT1; PICT-1; preS1 binding protein; PreS1-binding protein; Protein interacting with carboxyl terminus 1; R74911; Ribosome biogenesis protein NOP53
Gene Symbols NOP53
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human GLTSCR2. Recombinant protein control fragment (Product #RP-92581).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29997
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.