Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GLTSCR2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5113468
Description
GLTSCR2 Polyclonal Antibody for Western Blot, IHC (P), ELISA
Nucleolar protein which is involved in the integration of the 5S RNP into the ribosomal large subunit during ribosome biogenesis. In ribosome biogenesis, may also play a role in rRNA transcription. Also functions as a nucleolar sensor that regulates the activation of p53/TP53 in response to ribosome biogenesis perturbation, DNA damage and other stress conditions. DNA damage or perturbation of ribosome biogenesis disrupt the interaction between NOP53 and RPL11 allowing RPL11 transport to the nucleoplasm where it can inhibit MDM2 and allow p53/TP53 activation. It may also positively regulate the function of p53/TP53 in cell cycle arrest and apoptosis through direct interaction, preventing its MDM2-dependent ubiquitin-mediated proteasomal degradation. Originally identified as a tumor suppressor, it may also play a role in cell proliferation and apoptosis by positively regulating the stability of PTEN, thereby antagonizing the PI3K-AKT/PKB signaling pathway. May also inhibit cell proliferation and increase apoptosis through its interaction with NF2. May negatively regulate NPM1 by regulating its nucleoplasmic localization, oligomerization and ubiquitin-mediated proteasomal degradation. Thereby, may prevent NPM1 interaction with MYC and negatively regulate transcription mediated by the MYC-NPM1 complex. May also regulate cellular aerobic respiration. In the cellular response to viral infection, may play a role in the attenuation of interferon-beta through the inhibition of DDX58/RIG-1.
Specifications
GLTSCR2 | |
Polyclonal | |
Unconjugated | |
NOP53 | |
5330430H08Rik; 9430097C02Rik; AU041936; AW536441; glioma tumor suppressor candidate region gene 2; glioma tumor suppressor candidate region gene 2 protein; GLT; GLTSCR2; LOW QUALITY PROTEIN: glioma tumor suppressor candidate region gene 2 protein; NOP53; NOP53 ribosome biogenesis factor; p60; PICT1; PICT-1; preS1 binding protein; PreS1-binding protein; Protein interacting with carboxyl terminus 1; R74911; Ribosome biogenesis protein NOP53 | |
Rabbit | |
Protein G | |
RUO | |
292624, 29997 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
4.74 mg/mL | |
PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
Q9NZM5 | |
NOP53 | |
Recombinant Human Ribosome biogenesis protein NOP53 protein (227-405AA). | |
100 μg | |
Primary | |
Human, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction