Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Glucagon Antibody (09), mFluor Violet 500 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP221803MFV500
Description
Glucagon Monoclonal antibody specifically detects Glucagon in Human, Mouse, Rat, Porcine samples. It is validated for Flow CytometrySpecifications
Glucagon | |
Monoclonal | |
mFluor Violet 500 SE | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human, Mouse, Rat, Pig | |
Purified |
Flow Cytometry | |
09 | |
Flow Cytometry | |
glicentin-related polypeptide, GLP1, GLP-1, glucagon, glucagon-like peptide 1, glucagon-like peptide 2, GRPP | |
Synthetic full length oxyntomodulin coupled to carrier. | |
0.1 mL | |
Cancer, GPCR, Signal Transduction, Stem Cell Markers | |
2641 | |
Store at 4C in the dark. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction