Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Glucagon Antibody (49) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$206.50 - $499.50
Specifications
Antigen | GLP1 |
---|---|
Clone | 49 |
Concentration | 1 mg/mL |
Dilution | Western Blot 1:100-1:2000, ELISA 1:100-1:2000 |
Applications | Western Blot, ELISA |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB019350
![]() |
Novus Biologicals
NBP2235910.025ML |
0.025 mL |
Each for $206.50
|
|
|||||
NBP22359101
![]() |
Novus Biologicals
NBP2235910.1ML |
0.1 mL |
Each for $499.50
|
|
|||||
Description
Glucagon Monoclonal specifically detects Glucagon in Mammal samples. It is validated for Western Blot, ELISA.Specifications
GLP1 | |
1 mg/mL | |
Western Blot, ELISA | |
Unconjugated | |
Mouse | |
Mammalia, Human | |
glicentin-related polypeptide, GLP1, GLP2, glucagon, glucagon-like peptide 1, glucagon-like peptide 2, GRPP | |
GCG | |
IgG1 κ | |
Protein A or G purified | |
Clone 49 binds human GLP-1(9-36)amide Based on inhibition ELISA, Clone 49 cross-reacts 3% with GLP-1(7-36)amide |
49 | |
Western Blot 1:100-1:2000, ELISA 1:100-1:2000 | |
Monoclonal | |
Purified | |
Cancer, GPCR, Signal Transduction | |
10mM Phosphate (pH 7.4) and 0.5M NaCl with 15mM Sodium Azide | |
2641 | |
Synthetic GLP-1(9-18) adsorbed onto aluminum hydroxide gel | |
Primary | |
Store at 4C. Do not freeze. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title