Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Glucagon Antibody (53), Alexa Fluor™ 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP105184AF405
Description
Glucagon Monoclonal specifically detects Glucagon in Mammal samples. It is validated for Western Blot, ELISA.Specifications
Glucagon | |
Monoclonal | |
Alexa Fluor 405 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
GCG | |
Synthetic GLP-1(9-18) adsorbed onto aluminum hydroxide gel | |
0.1 mL | |
Cancer, GPCR, Signal Transduction | |
2641 | |
Human | |
Purified |
Western Blot, ELISA | |
53 | |
Western Blot, ELISA | |
glicentin-related polypeptide, GLP1, GLP-1, glucagon, glucagon-like peptide 1, glucagon-like peptide 2, GRPP | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Based on inhibition ELISA, It cross-reacts <2% with GLP-1(7-36)amide | |
Store at 4C in the dark. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction