Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glucagon Antibody (53) - BSA Free, Novus Biologicals™
SDP

Catalog No. NB018347 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
100 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB018347 0.025 mg
NBP105184 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB018347 Supplier Novus Biologicals Supplier No. NBP1051840.025MG
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Glucagon Monoclonal specifically detects Glucagon in Mammal samples. It is validated for Western Blot, ELISA.

Specifications

Antigen Glucagon
Applications ELISA, Western Blot
Classification Monoclonal
Clone 53
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, ELISA 1:6000
Formulation 10mM Phosphate (pH 7.4) and 0.5M NaCl with 0.05% Sodium Azide
Gene Alias glicentin-related polypeptide, GLP1, GLP-1, glucagon, glucagon-like peptide 1, glucagon-like peptide 2, GRPP
Gene Symbols GCG
Host Species Mouse
Immunogen Synthetic GLP-1(9-18) adsorbed onto aluminum hydroxide gel
Purification Method Protein A or G purified
Quantity 0.025 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2641
Test Specificity Based on inhibition ELISA, It cross-reacts <2% with GLP-1(7-36)amide
Target Species Human
Content And Storage Store at 4C. Do not freeze.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.