Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Glucagon Mouse anti-Human, Rat, Porcine, Clone: 8, Novus Biologicals™

Mouse Monoclonal Antibody
$206.50
Specifications
Antigen | Glucagon |
---|---|
Clone | 8 |
Concentration | 1 mg/ml |
Dilution | ELISA 1:100-1:2000, Sandwich ELISA |
Classification | Monoclonal |
Description
Glucagon Monoclonal specifically detects Glucagon in Human, Rat, Porcine samples. It is validated for ELISA, Sandwich ELISA.Specifications
Glucagon | |
1 mg/ml | |
Monoclonal | |
Purified | |
RUO | |
10mM Phosphate (pH 7.4) and 0.5M NaCl | |
2641 | |
IgG1 κ | |
Protein A or G purified |
8 | |
ELISA 1:100-1:2000, Sandwich ELISA | |
Unconjugated | |
Mouse | |
Cancer, GPCR, Signal Transduction, Stem Cell Markers | |
glicentin-related polypeptide, GLP1, GLP-1, glucagon, glucagon-like peptide 1, glucagon-like peptide 2, GRPP | |
Synthetic N-terminal fragment of glucagon coupled to carrier | |
Primary | |
Store at 4°C in the dark. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title