Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glucagon Mouse anti-Human, Unconjugated, Clone: 2A8G1, Proteintech
SDP

Catalog No. 502239413 Shop All Proteintech Group Inc Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-223-9413 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 50-223-9413 Supplier Proteintech Group Inc Supplier No. 672861IG150UL
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

The Glucagon antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human Glucagon. This antibody recognizes human, mouse antigen. The Glucagon antibody has been validated for the following applications: IF, IHC, ELISA analysis.

Specifications

Antigen Glucagon
Applications Immunohistochemistry, Immunofluorescence, ELISA
Classification Monoclonal
Clone 2A8G1
Concentration 1000 μg/mL
Conjugate Unconjugated
Host Species Mouse
Quantity 150 μL
Primary or Secondary Primary
Gene ID (Entrez) 2641
Target Species Human, Mouse
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.