Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Glucagon R/GCGR Antibody (591929) [DyLight 680], Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB10296Y
Description
Glucagon R/GCGR Monoclonal antibody specifically detects Glucagon R/GCGR in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, CyTOFSpecifications
Glucagon R/GCGR | |
Monoclonal | |
DyLight 680 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry, Immunohistochemistry, CyTOF | |
591929 | |
Flow Cytometry, Immunohistochemistry, CyTOF-ready | |
FLJ97182, GGR, GL-R, glucagon receptor, MGC138246 | |
HEK293 human embryonic kidney cell line transfected with human Glucagon R/GCGR, Met1-Gln142, Accession # P47871 | |
0.1 mL | |
GPCR, Signal Transduction | |
2642 | |
Store at 4°C in the dark. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction