Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Glucagon R/GCGR Antibody (hGR-2 F6), Alexa Fluor™ 532, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP281065AF532
Description
Glucagon R/GCGR Monoclonal specifically detects Glucagon R/GCGR in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
Glucagon R/GCGR | |
Monoclonal | |
Alexa Fluor 532 | |
50mM Sodium Borate | |
GCGR | |
Recombinant human Glucagon R/GCGR. | |
0.1 mL | |
GPCR, Signal Transduction | |
2642 | |
Human | |
Purified |
Immunofluorescence | |
hGR-2 F6 | |
Immunocytochemistry/Immunofluorescence | |
FLJ97182, GGR, GL-R, glucagon receptor, MGC138246 | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
The antibody binds to the human glucagon receptor, Glucagon R/GCGR. | |
Store at 4°C in the dark. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction