Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Glucagon R/GCGR Antibody (hGR-2 F6), Janelia Fluor™ 549, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP281065JF549
Description
Glucagon R/GCGR Monoclonal specifically detects Glucagon R/GCGR in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
| Glucagon R/GCGR | |
| Monoclonal | |
| Janelia Fluor 549 | |
| 50mM Sodium Borate | |
| GCGR | |
| Recombinant human Glucagon R/GCGR. | |
| 0.1 mL | |
| GPCR, Signal Transduction | |
| 2642 | |
| Human | |
| Purified |
| Immunofluorescence | |
| hGR-2 F6 | |
| Immunocytochemistry/Immunofluorescence | |
| FLJ97182, GGR, GL-R, glucagon receptor, MGC138246 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| The antibody binds to the human glucagon receptor, Glucagon R/GCGR. | |
| Store at 4°C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction