Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Glucagon R/GCGR Antibody (hGR-2 F6), PerCP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP281065PCP
Description
Glucagon R/GCGR Monoclonal antibody specifically detects Glucagon R/GCGR in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence, NeutralizationSpecifications
| Glucagon R/GCGR | |
| Monoclonal | |
| PerCP | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunocytochemistry, Neutralization | |
| hGR-2 F6 | |
| Immunocytochemistry/ Immunofluorescence, Block/Neutralize | |
| FLJ97182, GGR, GL-R, glucagon receptor, MGC138246 | |
| Recombinant human Glucagon R/GCGR. | |
| 0.1 mL | |
| GPCR, Signal Transduction | |
| 2642 | |
| Store at 4C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction