Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glucagon R/GCGR, Mouse anti-Human, Clone: hGR-2 F6, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

Supplier:  Novus Biologicals NBP2810650.2MG

 View more versions of this product

Catalog No. NB015225


Only null left
Add to Cart

Description

Description

Glucagon R/GCGR Monoclonal specifically detects Glucagon R/GCGR in Human samples. It is validated for Immunocytochemistry/Immunofluorescence, Block/Neutralize.
Specifications

Specifications

Glucagon R/GCGR
Monoclonal
1 mg/mL
Immunocytochemistry/Immunofluorescence, Block/Neutralize
FLJ97182, GGR, GL-R, glucagon receptor, MGC138246
Mouse
Protein A purified
RUO
Primary
The antibody binds to the human glucagon receptor, Glucagon R/GCGR.
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG1 κ
Immunofluorescence, Blocking Assay
hGR-2 F6
Unconjugated
PBS
GCGR
Recombinant human Glucagon R/GCGR.
0.2 mg
GPCR, Signal Transduction
2642
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.