Learn More
Glucagon Rabbit anti-Human, CoraLite 647, Polyclonal, Proteintech
Shop All Proteintech Group Inc Products

Description
The Glucagon antibody from Proteintech is a rabbit polyclonal antibody to a recombinant protein of human Glucagon. This antibody recognizes human, mouse, rat antigen. The Glucagon antibody has been validated for the following applications: IF analysis. This antibody is coralite™647.
Specifications
Specifications
| Antigen | Glucagon |
| Applications | Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | CoraLite 647 |
| Host Species | Rabbit |
| Quantity | 100 μL |
| Primary or Secondary | Primary |
| Gene ID (Entrez) | 2641 |
| Target Species | Human, Mouse, Rat |
| Isotype | IgG |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.