Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Glucokinase/GCK Antibody (OTI3E3), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP147778
Description
Glucokinase/GCK Monoclonal specifically detects Glucokinase/GCK in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
Glucokinase/GCK | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:500, Immunohistochemistry 1:50, Immunocytochemistry/Immunofluorescence 1:50, Immunohistochemistry-Paraffin 1:50 | |
GCK | |
Full length human recombinant protein of human GCK (NP_000153) produced in HEK293T cell. | |
Affinity Purified | |
RUO | |
2645 | |
Human, Mouse, Rat | |
IgG1 |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
OTI3E3 | |
Unconjugated | |
EC 2.7.1, EC 2.7.1.2, FGQTL3, GK, GLK, glucokinase (hexokinase 4), hexokinase-4, Hexokinase-D, HK4pancreatic isozyme, HKIV, HXKP, LGLK, maturity onset diabetes of the young 2, MODY2 | |
Mouse | |
52 kDa | |
0.1 mL | |
Primary | |
This antibody is specific for Homo sapiens glucokinase (hexokinase 4) (GCK), transcript variant 1. | |
Store at -20°C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction