Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Glucokinase/GCK Antibody (SR1917), Novus Biologicals™

Rabbit Monoclonal Antibody
$309.50 - $423.00
Specifications
Antigen | Glucokinase/GCK |
---|---|
Clone | SR1917 |
Dilution | Western Blot 1:500-1:2000 |
Applications | Western Blot |
Classification | Monoclonal |
Description
Glucokinase/GCK Monoclonal antibody specifically detects Glucokinase/GCK in Human samples. It is validated for Western BlotSpecifications
Glucokinase/GCK | |
Western Blot 1:500-1:2000 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
EC 2.7.1, EC 2.7.1.2, FGQTL3, GK, GLK, glucokinase (hexokinase 4), hexokinase-4, Hexokinase-D, HK4pancreatic isozyme, HKIV, HXKP, LGLK, maturity onset diabetes of the young 2, MODY2 | |
A synthesized peptide derived from human Glucokinase/GCK (Uniprot #: P35557) | |
Primary | |
Store at -20° C. Avoid freeze/thaw cycles. |
SR1917 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Lipid and Metabolism, Protein Kinase | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
2645 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title