Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Glucokinase/GCK, Mouse, Unlabeled, Clone: 9G1.2,
Mouse Monoclonal Antibody
Supplier: MilliporeSigma™ MABS1190
Description
Glucokinase (EC 2.7.1.2; UniProt P35557; also known as GK, Hexokinase-4, Hexokinase-D, Hexokinase type IV, HK IV, HK4) is encoded by the GCK (also known as HHF3, MODY2) gene (Gene ID 2645) in human. Glucokinase/GK functions as the body's primary glucose sensor by catalyzing the ATP-dependent phosphorylation of glucose to glucose-6-phosphate, the rate-limiting step of glucose metabolism in human pancreas and liver. GK activity is regulated rapidly in response to the elevated blood glucose after feeding, whereby a change in GK conformation causes a dissociation of the nuclear GK-glucokinase regulatory protein (GKRP) complex, allowing GK translocation to the cytoplasm in an activated state. In mammals, there exist four types of hexokinases, HK1, HK2, and HK3 have high affinity for glucose, while GK (also known as HK4) has low affinity for glucose and its expression is restricted to the pancreas and liver. GCK gene mutations cause Maturity-onset diabetes of the young 2 (MODY2) and Familial hyperinsulinemic hypoglycemia 3 (HHF3).
Specifications
Glucokinase/GCK | |
Monoclonal | |
Please refer to lot specific datasheet. | |
Purified mouse monoclonal IgG1κ antibody in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% Sodium Azide. | |
P35557 | |
GST-tagged recombinant internal fragment from the C-terminal half of human Glucokinase/GCK. | |
100 μg | |
Signaling | |
NP_000153 | |
Human | |
Purified |
Western Blot | |
9G1.2 | |
Unconjugated | |
GCK, HHF3, MODY2 | |
Mouse | |
Protein G Purified | |
RUO | |
Primary | |
Clone 9G1.2 recognizes an epitope present in all three spliced isoforms of human Glucokinase/GCK reported by UniProt (P35557). | |
Stable for 1 year at 2°-8°C from date of receipt. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only. Not for use in diagnostic procedures.
Spot an opportunity for improvement?Share a Content Correction