Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glucono delta-Lactone, FCC, 99-100.5%, Spectrum™ Chemical
SDP

Catalog No. 18602925 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
100 g
1 kg
5 kg
50 kg
Packaging:
Poly Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-602-925 1 kg Poly Bottle
18-602-924 100 g Poly Bottle
18-602-926 5 kg Poly Bottle
18-602-927 50 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-602-925 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. GL1071KG
Add to Cart
Add to Cart

GL107, 90-80-2, C6H10O6

Spectrum™ Chemical Glucono delta-Lactone, FCC is used as a sequestrant, a leavening agent, and an acidifier. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 90-80-2
Molecular Formula C6H10O6
Molecular Weight (g/mol) 178.14
MDL Number MFCD00006647
InChI Key PHOQVHQSTUBQQK-UHFFFAOYNA-N
IUPAC Name 3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-one
SMILES OCC1OC(=O)C(O)C(O)C1O

Specifications

CAS Max % 100%
Packaging Poly Bottle
Quantity 1 kg
Percent Purity 99 to 100.5%
Grade FCC
Physical Form Crystalline Powder or Lumps
Chemical Name or Material Glucono delta-Lactone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.