Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glucosamine Hydrochloride Dietary Supplement, USP, 98-102%, Spectrum™ Chemical
SDP

Catalog No. 18602936 Shop All Spectrum Chemical Manufacturing Corporation Products
Click to view available options
Quantity:
100 g
500 g
1 kg
5 kg
25 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail

Catalog No. 18-602-936

Supplier: Spectrum Chemical Manufacturing Corporation G109425KGBL

G1094, 66-84-2, C6H13NO5·HCl

Spectrum™ Chemical Glucosamine Hydrochloride, USP Dietary Supplement

Chemical Identifiers

CAS 66-84-2
Molecular Formula C6H14ClNO5
Molecular Weight (g/mol) 215.63
InChI Key CBOJBBMQJBVCMW-UHFFFAOYNA-N
IUPAC Name hydrogen 2-amino-3,4,5,6-tetrahydroxyhexanal chloride
SMILES [H+].[Cl-].NC(C=O)C(O)C(O)C(O)CO

Specifications

CAS Max % 100%
Ignition Residue 0.1%
Loss on Drying 1%
Packaging Fiber Drum
Quantity 25 kg
Specific Rotation +70.0° to +73.0
Percent Purity 98 to 102%
Grade USP Dietary Supplement
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.