Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Glucosamine (N-acetyl)-6-Sulfatase/GNS Antibody (007), DyLight 405, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289413V
Description
Glucosamine (N-acetyl)-6-Sulfatase/GNS Monoclonal specifically detects Glucosamine (N-acetyl)-6-Sulfatase/GNS in Human samples. It is validated for Western Blot, ELISA.Specifications
Glucosamine (N-acetyl)-6-Sulfatase/GNS | |
Monoclonal | |
DyLight 405 | |
EC 3.1.6, EC 3.1.6.14, G6Sglucosamine-6-sulfatase, glucosamine (N-acetyl)-6-sulfatase, Glucosamine-6-sulfatase, MGC21274, N-acetylglucosamine-6-sulfatase | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human Glucosamine (N-acetyl)-6-Sulfatase/GNS (NP_002067.1; Met 1-Leu 552). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot, ELISA | |
7 | |
50 mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
2799 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction