Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glucosamine Sulfate Potassium Chloride, Dietary Supplement, USP, 98-102%, Spectrum™ Chemical
SDP

Catalog No. 18602937 Shop All Spectrum Chemical Manufacturing Corporation Products
Click to view available options
Quantity:
100 g
12 kg
25 kg
Packaging:
Poly Bottle
Poly Pail

Catalog No. 18-602-937

Supplier: Spectrum Chemical Manufacturing Corporation G1296100GM

G1296, 1296149-08-0, (C6H14NO5)2SO4·2KCl

Spectrum™ Chemical Glucosamine Sulfate Potassium Chloride, Dietary Supplement, USP slows deterioration of cartilage, relieves osteoarthritis (OA) pain and improves joint mobility. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 1296149-08-0

Specifications

CAS Max % 100%
Ignition Residue 26.5-31%
Loss on Drying 1%
Packaging Poly Bottle
Quantity 100 g
Specific Rotation 47.0° to +53.0
Percent Purity 98 to 102%
Grade USP
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.