Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Glucosaminyl (N-acetyl) Transferase 1/GCNT1 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$181.90 - $487.50
Specifications
Antigen | Glucosaminyl (N-acetyl) Transferase 1/GCNT1 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Description
Glucosaminyl (N-acetyl) Transferase 1/GCNT1 Polyclonal specifically detects Glucosaminyl (N-acetyl) Transferase 1/GCNT1 in Human samples. It is validated for Western Blot.Specifications
Glucosaminyl (N-acetyl) Transferase 1/GCNT1 | |
Polyclonal | |
Purified | |
RUO | |
PBS with 50% glycerol, pH7.3. | |
2650 | |
IgG | |
Affinity purified |
Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
beta-1,3-galactosyl-O-glycosyl-glycoproteinbeta-1,6-N-acetylglucosaminyltransferase, beta-1,6-N-acetylglucosaminyltransferase, C2GNT1, C2GNTbeta-1,3-galactosyl-O-glycosyl-glycoproteinbeta-1,6-N--acetylglucosaminyltransferase, C2GNT-L, core 2 beta1,6 N-acetylglucosaminyltransferase-I, core 2 beta-1,6-N-acetylglucosaminyltransferase I, core 2 branching enzyme, Core 2 GNT, Core 2-branching enzyme, Core2-GlcNAc-transferase, EC 2.4.1.102, G6NT, glucosaminyl (N-acetyl) transferase 1, core 2, glucosaminyl (N-acetyl) transferase 1, core 2(beta-1,6-N-acetylglucosaminyltransferase), MGC126335, MGC126336, NACGT2, NAGCT2 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 40-300 of human Glucosaminyl (N-acetyl) Transferase 1/GCNT1 (NP_001091103.1). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title