Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Glucose 1-dehydrogenase Antibody (OTI2A7), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271478F
Description
Glucose 1-dehydrogenase Monoclonal specifically detects Glucose 1-dehydrogenase in Human, Monkey, Primate samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
Glucose 1-dehydrogenase | |
Monoclonal | |
FITC | |
PBS with 0.05% Sodium Azide | |
H6PD | |
Full length recombinant protein of human Glucose 1-dehydrogenase (NP_004276) produced in HEK293T cells. | |
0.1 mL | |
9563.0 | |
Store at 4°C in the dark. |
Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
OTI2A7 | |
Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
6-phosphogluconolactonase, DKFZp686A01246, EC 1.1.1.49, EC 2.7.4.3, EC 3.1.1.31, G6PD, H form, G6PDH, GDH/6PGL endoplasmic bifunctional protein, GDHglucose 1- dehydrogenase, glucose dehydrogenase, glucose dehyrogenase, glucose-6-phosphate dehydrogenase, salivary, hexose-6-phosphate dehydrogenase (glucose 1-dehydrogenase), MGC87643 | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Monkey, Primate | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction