Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glucose 6 phosphatase 2 Polyclonal Antibody, Biotin, Bioss
SDP

Catalog No. 502280507
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-228-0507 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 50-228-0507 Supplier Bioss Supplier No. BS22837RBIOTIN
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Glucose 6 phosphatase 2 Polyclonal antibody specifically detects Glucose 6 phosphatase 2 in Human, Mouse, Rat samples. It is validated for Western Blot

Specifications

Antigen Glucose 6 phosphatase 2
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Biotin
Formulation TBS with 1% BSA, 50% glycerol and 0.03% ProClin 300
Gene G6PC2
Gene Accession No. Q9NQR9, Q9Z186
Gene Alias G6Pase 2; G-6-Pase 2; G6PC2; G6pc-rs; Glucose-6-phosphatase 2; glucose-6-phosphatase catalytic subunit 2; glucose-6-phosphatase, catalytic, 2; glucose-6-phosphatase, catalytic, related sequence; IGRP; islet-specific G6CP-related protein; islet-specific glucose-6-phosphatase catalytic subunit-related protein; islet-specific glucose-6-phosphatase-related protein; LOC681817; similar to glucose-6-phosphatase, catalytic, related
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human Glucose 6 phosphatase 2 (191-290).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14378, 57818, 681817
Target Species Human, Mouse, Rat
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.