Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Glucose 6 phosphatase 2 Polyclonal Antibody, HRP, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS22837RHRP

This item is not returnable.
View return policy
Description
Glucose 6 phosphatase 2 Polyclonal antibody specifically detects Glucose 6 phosphatase 2 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
Glucose 6 phosphatase 2 | |
Polyclonal | |
HRP | |
G6PC2 | |
G6Pase 2; G-6-Pase 2; G6PC2; G6pc-rs; Glucose-6-phosphatase 2; glucose-6-phosphatase catalytic subunit 2; glucose-6-phosphatase, catalytic, 2; glucose-6-phosphatase, catalytic, related sequence; IGRP; islet-specific G6CP-related protein; islet-specific glucose-6-phosphatase catalytic subunit-related protein; islet-specific glucose-6-phosphatase-related protein; LOC681817; similar to glucose-6-phosphatase, catalytic, related | |
KLH conjugated synthetic peptide derived from human Glucose 6 phosphatase 2 (191-290). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Western Blot | |
1 mg/mL | |
TBS with 1% BSA, 50% glycerol and 0.03% ProClin 300 | |
Q9NQR9, Q9Z186 | |
Rabbit | |
Protein A | |
RUO | |
14378, 57818, 681817 | |
-20°C or -80°C if preferred | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction