Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glucose 6 Phosphate Dehydrogenase Antibody (2H7), Allophycocyanin, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

Supplier:  Novus Biologicals NBP222125APC

Catalog No. NB222125APC


Only null left
Add to Cart

Description

Description

Glucose 6 Phosphate Dehydrogenase Monoclonal specifically detects Glucose 6 Phosphate Dehydrogenase in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

Glucose 6 Phosphate Dehydrogenase
Monoclonal
APC
PBS with 0.05% Sodium Azide
G6PD
Purified recombinant fragment of human Glucose 6 Phosphate Dehydrogenase expressed in E. coli. [Uniprot: P11413]
0.1 mL
Cancer, Cellular Markers, Golgi Apparatus Markers
2539
Store at 4C in the dark.
IgG1
Flow Cytometry
2H7
Flow Cytometry
G6PD, G6PD1EC 1.1.1.49, glucose-6-phosphate dehydrogenase
Mouse
Ammonium sulfate precipitation
RUO
Primary
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.

For Research Use Only