Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Glucose 6 Phosphate Dehydrogenase Antibody (2H7), Biotin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP222125B
Description
Glucose 6 Phosphate Dehydrogenase Monoclonal specifically detects Glucose 6 Phosphate Dehydrogenase in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Glucose 6 Phosphate Dehydrogenase | |
Monoclonal | |
Biotin | |
PBS with 0.05% Sodium Azide | |
G6PD | |
Purified recombinant fragment of human Glucose 6 Phosphate Dehydrogenase expressed in E. coli. [Uniprot: P11413] | |
0.1 mL | |
Cancer, Cellular Markers, Golgi Apparatus Markers | |
2539 | |
Store at 4C in the dark. | |
IgG1 |
Western Blot, ELISA, Immunohistochemistry (Paraffin) | |
2H7 | |
Western Blot, ELISA, Immunohistochemistry-Paraffin | |
G6PD, G6PD1EC 1.1.1.49, glucose-6-phosphate dehydrogenase | |
Mouse | |
Ammonium sulfate precipitation | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction