Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Glucose 6 Phosphate Dehydrogenase Antibody, HRP, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB100236H
Description
Glucose 6 Phosphate Dehydrogenase Polyclonal specifically detects Glucose 6 Phosphate Dehydrogenase in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunoblotting.Specifications
Glucose 6 Phosphate Dehydrogenase | |
Polyclonal | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunoblotting | |
G6PD, G6PD1EC 1.1.1.49, glucose-6-phosphate dehydrogenase | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Human, Mouse | |
IgG |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunoblot | |
HRP | |
PBS with No Preservative | |
G6PD | |
The immunogen recognized by this antibody maps to a region between residues 50 and 100 of human Glucose-6-Phosphate Dehydrogenase using the numbering given in entry NP_000393.2 (GeneID 2539). | |
100 μL | |
Cancer, Cellular Markers, Golgi Apparatus Markers | |
2539 | |
Store at 4C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction