Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Glucose 6 phosphate isomerase Rabbit anti-Human, Rat, Clone: 2Y8R0, Novus Biologicals™

Rabbit Monoclonal Antibody
$205.50 - $458.50
Specifications
Antigen | Glucose 6 phosphate isomerase |
---|---|
Clone | 2Y8R0 |
Applications | Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
Glucose 6 phosphate isomerase Monoclonal antibody specifically detects Glucose 6 phosphate isomerase in Human, Rat samples. It is validated for Western BlotSpecifications
Glucose 6 phosphate isomerase | |
Western Blot | |
Unconjugated | |
Rabbit | |
Cytokine Research | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
2821 | |
IgG | |
Affinity purified |
2Y8R0 | |
Monoclonal | |
Purified | |
RUO | |
Human, Rat | |
AMFGNPI, Autocrine motility factor, DKFZp686C13233, EC 5.3.1.9, glucose phosphate isomerase, glucose-6-phosphate isomerase, hexose monophosphate isomerase, hexosephosphate isomerase, neuroleukin, NLKSA36, oxoisomerase, PGI, PHI, Phosphoglucose isomerase, phosphohexomutase, Phosphohexose isomerase, phosphosaccharomutase, SA-36, Sperm antigen 36, sperm antigen-36 | |
A synthetic peptide corresponding to a sequence within amino acids 1-100 of human Glucose 6 phosphate isomerase (P06744). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title