Learn More
Description
Includes
- Glucose Oxidase >15u/ml; Peroxidase (horseradish) 800U/L; 4-
- Aminoantipyrine 0.38mM; Phosphate Buffer; pH 7.5±0.1; Sodium p-Hydroxybenzoate 5mM; non-reactive Stabilizers and fillers; Sodium Azide 0.1%
- Method: Oxidase/Trinder, Endpoint
- Format: Dry powder
- Wavelength: 500nm
- Linearity: 500mg/dL
- Expected Values: 70 to 105mg/dL
- Reconstituted stability: 30 days at 2 to 8°C
For the quantitative determination of glucose in serum
Specifications
Specifications
| Certifications/Compliance | CE Marked |
| Type | Glucose (Oxidase) Reagent Kit |
| Formulation | Glucose Oxidase (microbial) 12000U/L, Peroxidase (horseradish) >1000U/L,4-AAP >0.3mM, Phenol 4mM, Buffer pH 7.4 ± 0.1, non-reactive Stabilizers |
| For Use With (Application) | Quantitative determination of Glucose in Serum, in-vitro diagnostic use only |
| pH | 7.3 |
| Quantity | 10 x 100mL (3.4 oz.) |
| Form | Liquid |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.