Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega Glucose-Glo Assay, 50ml

Catalog No. PRJ6022 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
50 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRJ6022 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PRJ6022 Supplier Promega Supplier No. J6022
May include imposed supplier surcharges.
Add to Cart
Add to Cart

A bioluminescent assay for rapid and sensitive measurement of glucose from a variety of sample types. The assay is suitable for detecting altered glucose consumption due to changes in glycolysis or glucose production during gluconeogenesis.

  • Detect Even Small Changes in Glucose
  • Measure glucose in a variety of sample types
  • Limit of detection down to nM range
  • Signal to background >1,000

Specifications

Content And Storage LESS THAN -65°C
Quantity 50 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.